Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCSAML Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565012
Description
Immunogen sequence: CYTVINHIPHQ RSSLSSNDDG YENIDSLTRK VRQFRERSET EYALLRTSVS RPCSCTHEHD YEVVF Highest antigen sequence identity to the following orthologs - mouse 29%, rat 32%.
This gene encodes a protein thought to be a signaling molecule associated with germinal centers, the sites of proliferation and differentiation of mature B lymphocytes. Alternate splicing results in multiple transcript variants.
Specifications
GCSAML | |
Polyclonal | |
Unconjugated | |
GCSAML | |
C1orf150; GCSAML; germinal center associated signaling and motility like; germinal center-associated signaling and motility-like protein; germinal center-associated, signaling and motility-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
148823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JQS6 | |
GCSAML | |
Recombinant Human GCSAML. Recombinant protein control fragment (Product #RP-105777). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction