Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCSH Antibody (105), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290289PCP
Description
GCSH Monoclonal antibody specifically detects GCSH in Human samples. It is validated for ELISASpecifications
GCSH | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
105 | |
ELISA | |
GCE, glycine cleavage system H protein, mitochondrial, glycine cleavage system protein H (aminomethyl carrier), lipoic acid-containing protein, mitochondrial glycine cleavage system H-protein, NKH | |
Rabbit immunized with purified, recombinant Human GCSH (Accession#: AAA36011.1, Ser49-Glu173) | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
2653 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction