Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCSH Antibody (187), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290290AF532
Description
GCSH Monoclonal specifically detects GCSH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GCSH | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2653 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
187 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
GCE, glycine cleavage system H protein, mitochondrial, glycine cleavage system protein H (aminomethyl carrier), lipoic acid-containing protein, mitochondrial glycine cleavage system H-protein, NKH | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human GCSH (AAA36011.1; Ser49-Glu173). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction