Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCSH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559538
Description
Immunogen sequence: TLRTGPALLS VRKFTEKHEW VTTENGIGTV GISNFAQEAL GDVVYCSLPE VGTKLNKQDE FGA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
The glycine cleavage system catalyzes the degradation of glycine. The H protein (GCSH) shuttles the methylamine group of glycine from the P protein (GLDC) to the T protein (GCST).
Specifications
GCSH | |
Polyclonal | |
Unconjugated | |
GCSH | |
1100001L02Rik; 5730591C18Rik; GCE; Gcsh; Glycine cleavage system H protein, mitochondrial; glycine cleavage system protein H; glycine cleavage system protein H (aminomethyl carrier); lipoic acid-containing protein; mitochondrial glycine cleavage system H-protein; NKH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171133, 2653, 68133 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23434, Q5I0P2, Q91WK5 | |
GCSH | |
Recombinant protein corresponding to Human GCSH. Recombinant protein control fragment (Product #RP-98558). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction