Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCX1/C20orf100 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981337
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1337 Supplier Bioss Supplier No. BS11402R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOX2 (TOX High Mobility Group Box Family Member 2) is a Protein Coding gene. Diseases associated with TOX2 include Conduct Disorder. Gene Ontology (GO) annotations related to this gene include chromatin binding and RNA polymerase II transcription factor binding. An important paralog of this gene is TOX.

Specifications

Antigen GCX1/C20orf100
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TOX2
Gene Accession No. Q96NM4
Gene Alias AI851523; AV026525; BI987407; C20orf100; dJ1108D11.2; dJ495O3.1; GCX1; Gcx-1; granulosa cell HMG box 1; Granulosa cell HMG box protein 1; granulosa cell HMG-box protein 1; granulosa cell HMG-box protein-1; LOC100347442; RxHMG1; TOX high mobility group box family member 2; TOX2
Gene Symbols TOX2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GCX1/C20orf100.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84969
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.