Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121962
This item is not returnable.
View return policy
Description
GDA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
Specifications
GDA | |
Polyclonal | |
Unconjugated | |
GDA | |
AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9615 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Y2T3 | |
GDA | |
Recombinant protein Guanine Deaminase. The antigen corresponds to amino acid range 1-454 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction