Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579304
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579304 Supplier Invitrogen™ Supplier No. PA579304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, rat liver tissue, rat small intestine tissue, mouse brain tissue, mouse liver tissue, mouse small intestine tissue. IHC: human colon cancer tissue, mouse small intestine tissue, rat small intestine tissue.

GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene GDA
Gene Accession No. Q9R111, Q9WTT6, Q9Y2T3
Gene Alias AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin
Gene Symbols GDA
Host Species Rabbit
Immunogen E. coli-derived human GDA recombinant protein (Position: E220-Q446).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14544, 83585, 9615
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.