Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553136 Supplier Invitrogen™ Supplier No. PA553136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLSEHNEPWF MRLNSTGEVP VLIHGENIIC EATQIIDYLE QTFLDERTPR LMPDKESMYY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 97%.

Glutathione S-transferases (GSTs) function to conjugate reduced glutathione to many exogenous and endogenous hydrophobic electrophiles. Although it shares the carboxy and amino-terminal glutathione S-transferase domains, GDAP1 (ganglioside-induced differentiation-associated protein 1) is characterized as a GST-like protein because it contains an extended GST domain II and a predicted transmembrane domain, two characteristics which are unusual for GST family members. GDAP1 may function in a signal transduction pathway that is responsible for ganglioside-induced neurite differentiation and also may play a role in protecting myelin membranes from free-radical damage. Mutations in the gene encoding GDAP1 is the cause of many forms of Charcot-Marie-Tooth disease, a common inherited disorder of the peripheral nervous system that is characterized by reduced nerve conduction velocities, slow progressive distal muscle atrophy and absent deep tendon reflexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GDAP1
Gene Accession No. Q8TB36
Gene Alias Charcot-Marie-Tooth neuropathy 4A; CMT4; CMT4A; CMTRIA; ganglioside induced differentiation associated protein 1; ganglioside-induced differentiation-associated protein 1; ganglioside-induced differentiation-associated-protein 1; Gdap1
Gene Symbols GDAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GDAP1. Recombinant protein control fragment (Product #RP-90927).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.