Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDAP1L1 Antibody (OTI1G5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272182AF532
Description
GDAP1L1 Monoclonal specifically detects GDAP1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
GDAP1L1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
GDAP1L1 | |
Full length recombinant protein of human GDAP1L1 (NP_076939) produced in HEK293T cells. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1G5 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
dJ881L22.1, dJ995J12.1.1, DKFZp761K228, ganglioside-induced differentiation-associated protein 1-like 1, GDAP1-L1 | |
Mouse | |
0.1 mL | |
78997 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction