Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDAP1L1 Antibody (OTI1G5), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB275811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB275811 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB275811 Supplier Novus Biologicals Supplier No. NBP272182MFV500
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GDAP1L1 Monoclonal antibody specifically detects GDAP1L1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen GDAP1L1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1G5
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias dJ881L22.1, dJ995J12.1.1, DKFZp761K228, ganglioside-induced differentiation-associated protein 1-like 1, GDAP1-L1
Host Species Mouse
Immunogen Full length human recombinant protein of human GDAP1L1(NP_076939) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78997
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.