Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDAP1L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569628
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. Several transcript variants encoding different isoforms, as well as a noncoding transcript variant, have been found for this gene.
Specifications
GDAP1L1 | |
Polyclonal | |
Unconjugated | |
GDAP1L1 | |
dJ881L22.1; dJ995J12.1.1; ganglioside induced differentiation associated protein 1 like 1; ganglioside induced differentiation associated protein 1-like 1; ganglioside induced differentiation associated protein 1-like 1; ganglioside-induced differentiation-associated protein 1-like 1; ganglioside-induced differentiation-associated protein 1-like 1; Gdap1l; GDAP1L1; GDAP1-L1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
311616 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
GDAP1L1 | |
synthetic peptide directed towards the C-terminal region of rat GDAP1L1. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction