Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    GDAP1L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 119771AP150UL
 
            Description
The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. Several transcript variants encoding different isoforms, as well as a noncoding transcript variant, have been found for this gene.Specifications
| GDAP1L1 | |
| Polyclonal | |
| Unconjugated | |
| GDAP1L1 | |
| GDAP1 L1, GDAP1L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 228858, 78997 | |
| -20°C | |
| Liquid | 
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VE33, Q96MZ0 | |
| GDAP1L1 | |
| GDAP1L1 Fusion Protein Ag2602 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                             
                             
                             
                            