Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561175
Description
Immunogen sequence: LNSSDTTAEI FQEDTVRSPF LYNKDVNGKV VLWKGDVALL NCTAIVNTSN ESLTDKNPVS ESIFMLAGPD LKEDLQKLKG CRTGEAKL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 92%.
GDAP2 is a protein coding gene and gene ontology (GO) annotation includes lysosomal membrane; response to retinoic acid.
Specifications
GDAP2 | |
Polyclonal | |
Unconjugated | |
GDAP2 | |
C77050; D3Ertd801e; dJ776P7.1; ganglioside induced differentiation associated protein 2; ganglioside-induced differentiation-associated protein 2; Ganglioside-induced differentiation-associated-protein 2; GDAP2; MACROD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54834 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXN4 | |
GDAP2 | |
Recombinant protein corresponding to Human GDAP2. Recombinant protein control fragment (Product #RP-94543). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction