Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GDC 0879
GREENER_CHOICE

Catalog No. 445310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-531-0 10 mg
44-535-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-531-0 Supplier Tocris Bioscience™ Supplier No. 4453/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent B-Raf inhibitor

Potent and selective B-Raf inhibitor (IC50 = 0.13 nM against purified B-Raf V600E). Activity reduces phospho-ERK (pERK) levels (IC50 = 63 nM in the Malme-3M cell line). Inhibits the Raf/MEK/ERK signaling pathway in V600E B-Raf mutant cell lines. Does not activate apoptotic pathways in A375 or Colo205 cell lines. Orally bioavailable.

Chemical Identifiers

CAS 905281-76-7
Molecular Formula C19H18N4O2

Specifications

Quantity 10 mg
Formula Weight 334.37
Purity >98%
Molecular Weight (g/mol) 334.379
Chemical Name or Material GDC 0879
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.