Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | GDC |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GDC Polyclonal specifically detects GDC in Human samples. It is validated for Western Blot.Specifications
GDC | |
Polyclonal | |
Rabbit | |
P16260 | |
8034 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
D10S105EGDC, GDASolute carrier family 25 member 16, Graves disease autoantigen, graves disease carrier protein, HGT.1, hML7, member 16, MGC39851, Mitochondrial solute carrier protein homolog, ML7, solute carrier family 25 (mitochondrial carrier; Graves disease autoantigen) | |
SLC25A16 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title