Learn More
Invitrogen™ GDE1 Monoclonal Antibody (2H6)

Description
Peptide Sequence: ACLLTGSLFV LLRVFSFEPV PSCRALQVLK PRDRISAIAH RGGSHDAPEN TLAAIRQAAK NGATGVELDI EFTSDGIPVL MHDNTVDRTT DGTGRLCDLT FEQIRKLN.
Specifications
Specifications
| Antigen | GDE1 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2H6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Gde1 |
| Gene Accession No. | Q9NZC3 |
| Gene Alias | 1200003M13Rik; 363E6.2; Gde1; Glycerophosphodiester phosphodiesterase 1; Glycerophosphodiester phosphodiesterase 1-like protein; hypothetical protein LOC539978; membrane interacting protein of RGS16; membrane-interacting protein of RGS16; MIR16; PAMCI; PCIP1; RGS16; RGS16-interacting membrane protein; similar to membrane interacting protein of RGS16; zgc:56068; zgc:56068 protein; zgc:77135 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.