Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143255
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143255 Supplier Invitrogen™ Supplier No. PA5143255
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Has glycerophosphoinositol phosphodiesterase activity. Has little or no activity towards glycerophosphocholine. GDE1 activity can be modulated by G-protein signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDE1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Gde1
Gene Accession No. Q9JL55, Q9JL56
Gene Alias 1200003M13Rik; 363E6.2; Gde1; Glycerophosphodiester phosphodiesterase 1; Glycerophosphodiester phosphodiesterase 1-like protein; hypothetical protein LOC539978; membrane interacting protein of RGS16; membrane-interacting protein of RGS16; MIR16; PAMCI; PCIP1; RGS16; RGS16-interacting membrane protein; similar to membrane interacting protein of RGS16; zgc:56068; zgc:56068 protein; zgc:77135
Gene Symbols Gde1
Host Species Goat
Immunogen Peptide with sequence C-SLSHTGDGKPRYD (aa 230-242).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56209, 60418
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.