Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-15 Antibody (7C12.B3.F2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142725AF700
Description
GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Western Blot, ELISA.Specifications
GDF-15 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
7C12.B3.F2 | |
Western Blot, ELISA | |
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
This Protein A purified GDF-15 Antibody (7C12.B3.F2) was prepared by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminal end of human GDF-15 protein. A residue of cysteine was added to facilitate coupling to KLH. (Uniprot: Q99988) | |
0.1 mL | |
Hepatic Stem Cell Markers, Stem Cell Markers | |
9518 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction