Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-15 Antibody (MM0307-10A3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | GDF-15 |
---|---|
Clone | MM0307-10A3 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
GDF-15 | |
LYOPH | |
Unconjugated | |
Mouse | |
Human | |
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
GDF15 | |
Protein G purified | |
This antibody was selected for its ability to detect human GDF-15. |
MM0307-10A3 | |
Monoclonal | |
Purified | |
RUO | |
Q99988.2 | |
9518 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title