Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-15 Antibody (MM0307-10A3), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212331PE
Description
GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Flow Cytometry.Specifications
GDF-15 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
GDF15 | |
Human recombinant GDF-15 | |
0.1 mL | |
Primary | |
This antibody was selected for its ability to detect human GDF-15. | |
Store at 4C in the dark. | |
IgG2 |
Flow Cytometry | |
MM0307-10A3 | |
Flow Cytometry | |
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
Mouse | |
Protein G purified | |
Hepatic Stem Cell Markers, Stem Cell Markers | |
9518 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction