Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-5/BMP-14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | GDF-5/BMP-14 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GDF-5/BMP-14 Polyclonal specifically detects GDF-5/BMP-14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GDF-5/BMP-14 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
BMP14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 | |
GDF5 | |
IgG | |
Affinity Purified | |
Specificity of human GDF-5/BMP-14 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Rabbit | |
Cytokine Research | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
8200 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RYVFDISALEKDGLLGAELRILRKKPSDTAKPAAPGGGRAAQLKLSSCPSGRQPASLLDVRSVPGLDGSGWEVFDIWKLFRNFKNSAQLCLELEAWERGRAVDL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title