Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-5/BMP-14 Antibody (RM0242-13J15), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212333AF350
Description
GDF-5/BMP-14 Monoclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot.Specifications
GDF-5/BMP-14 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
GDF5 | |
Mouse recombinant GDF-5/BMP-14 | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Western Blot | |
RM0242-13J15 | |
Western Blot | |
BMP14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 | |
Rat | |
Protein A or G purified | |
Cytokine Research | |
8200.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction