Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-5/BMP-14 Antibody (RM0242-13J15) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen GDF-5/BMP-14
Clone RM0242-13J15
Concentration LYOPH
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019176
SDP
View Documents
Novus Biologicals
NBP2123330.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP212333
SDP
View Documents
Novus Biologicals
NBP212333
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

GDF-5/BMP-14 Monoclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

GDF-5/BMP-14
LYOPH
Unconjugated
Rat
Cytokine Research
P43027
8200
IgG2
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
RM0242-13J15
Monoclonal
Purified
RUO
Mouse
BMP14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2
GDF5
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.