Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-8/Myostatin Antibody (6E4E6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$541.95
Specifications
| Antigen | GDF-8/Myostatin |
|---|---|
| Clone | 6E4E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
GDF-8/Myostatin Monoclonal specifically detects GDF-8/Myostatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GDF-8/Myostatin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
| MSTN | |
| IgG2b | |
| Affinity Purified |
| 6E4E6 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Cytokine Research | |
| PBS with 0.05% Sodium Azide | |
| 2660 | |
| Purified recombinant fragment of human MSTN (AA:24-266) expressed in E. Coli. | |
| Primary | |
| 42.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title