Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-8/Myostatin Antibody (GDF9/4261), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308680AF405
Description
GDF-8/Myostatin Monoclonal antibody specifically detects GDF-8/Myostatin in Human samples. It is validated for Western Blot, ELISASpecifications
GDF-8/Myostatin | |
Monoclonal | |
Alexa Fluor 405 | |
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
Recognizes the 113 amino acid carboxy-terminal fragment of GDF-8/Myostatin protein (Uniprot: O14793) | |
100 μg | |
Cytokine Research | |
2660 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
GDF9/4261 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction