Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-8/Myostatin Antibody (OTI7H5), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270826AF594
Description
GDF-8/Myostatin Monoclonal specifically detects GDF-8/Myostatin in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GDF-8/Myostatin | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MSTN | |
Recombinant protein fragment corresponding to amino acids 24-266 of human GDF-8/Myostatin (NP_005250) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI7H5 | |
Western Blot | |
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
Mouse | |
Affinity Purified | |
Cytokine Research | |
2660.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction