Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-8/Myostatin Antibody (OTI7H5) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270826
Description
GDF-8/Myostatin Monoclonal specifically detects GDF-8/Myostatin in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GDF-8/Myostatin | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
Mouse | |
42.6 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI7H5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MSTN | |
Human recombinant protein fragment corresponding to amino acids 24-266 of human MSTN(NP_005250) produced in E.coli. | |
Affinity Purified | |
Cytokine Research | |
2660 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction