Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-8/Myostatin Antibody (RM0243-13F2) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$192.47
Specifications
| Antigen | GDF-8/Myostatin |
|---|---|
| Clone | RM0243-13F2 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
Description
GDF-8/Myostatin Monoclonal specifically detects GDF-8/Myostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GDF-8/Myostatin | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
| MSTN | |
| Primary |
| RM0243-13F2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rat | |
| Cytokine Research | |
| O08689 | |
| 2660 | |
| IgG2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title