Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-9 Antibody (53/1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261934R
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
GDF-9 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GDF9 | |
Tuberculin coupled peptide with sequence VPAKYSPLSVLTIEPDGSIAYKEYEDMIATKC that recognizes an epitope with the EPDG sequence near the C-terminal region of human GDF-9 | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Western Blot | |
53/1 | |
Western Blot, ELISA, Immunohistochemistry | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
Mouse | |
Protein A purified | |
Cytokine Research | |
2661 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction