Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-9 Antibody (72B), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP261933G
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA.Specifications
GDF-9 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GDF9 | |
N-terminal of GDF-9 corresponding to sequence KKPLGPASFNLSEYFC | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Western Blot | |
72B | |
Western Blot, ELISA | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
Rat | |
Protein G purified | |
Cytokine Research | |
2661 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction