Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-9 Antibody (917319) [Allophycocyanin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8266A
Description
GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
GDF-9 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
917319 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
Human embryonic kidney cell line derived recombinant human GDF-9, Met1-Arg454, Accession # O60383 | |
0.1 mL | |
Cytokine Research | |
2661 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction