Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-9 Rat anti-Human, Clone: GDF9/4262, Novus Biologicals™

Rat Monoclonal Antibody
$268.00
Specifications
| Antigen | GDF-9 |
|---|---|
| Clone | GDF9/4262 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, ELISA |
| Applications | Western Blot, ELISA |
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA.Specifications
| GDF-9 | |
| 0.2 mg/ml | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Cytokine Research | |
| 10 mM PBS with 0.05% BSA | |
| 2661 | |
| IgG1 | |
| Protein A or G purified |
| GDF9/4262 | |
| Western Blot 1-2 ug/ml, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
| N-terminal of GDF-9, corresponding to sequence KKPLGPASFNLSEYFC. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title