Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119164
This item is not returnable.
View return policy
Description
GDF1 Polyclonal Antibody for Western Blot, IHC (P)
Growth differentiation factors (GDFs) are members of the transforming growth factor (TGF) superfamily that is involved in embryonic development and adult tissue homeostasis. GDF1 was initially identified as a temporally expressed gene in the mouse central nervous system during embryonic development, with only one isoform detected in adult tissues. GDF1 is required for left-right patterning during development and directly interacts with Nodal, another member of the TGF-beta superfamily. It has been suggested that GDF1 regulates the activity and signaling range of Nodal through direct interaction.
Specifications
GDF1 | |
Polyclonal | |
Unconjugated | |
GDF1 | |
AI385651; DORV; DTGA3; Embryonic growth/differentiation factor 1; Gdf1; GDF-1; growth differentiation factor 1; RAI | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
14559 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P20863 | |
GDF1 | |
Recombinant protein Growth Differentiation Factor 1. The antigen corresponds to amino acid range 230-357 of the target protein. | |
200 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction