Learn More
Invitrogen™ GDF10 Polyclonal Antibody

Description
GDF10 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | GDF10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GDF10 |
| Gene Accession No. | P55107 |
| Gene Alias | BIP; Bmp3b; BMP-3B; Bone morphogenetic protein 3B; bone morphogenetic protein 3B; growth/differentiation factor 10; bone-inducing protein; Gdf10; GDF-10; growth differentiation factor 10; growth/differentiation factor 10; prepro bone inducing protein |
| Gene Symbols | GDF10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.