Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF11 Monoclonal Antibody (1956CT308.38.25)
GREENER_CHOICE

Catalog No. PIMA537765
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537765 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537765 Supplier Thermo Scientific Supplier No. MA537765
Add to Cart
Add to Cart

GDF11 Monoclonal Antibody (1956CT308.38.25)

GDF11 Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF11
Applications Western Blot
Classification Monoclonal
Clone 1956CT308.38.25
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GDF11
Gene Accession No. O95390
Gene Alias BMP11; BMP-11; bone morphogenetic protein 11; GDF11; GDF-11; growth differentiation factor 11; growth/differentiation factor 11
Gene Symbols GDF11
Host Species Mouse
Immunogen recombinant protein from the human region of human GDF11.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.