Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDF15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547839
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547839 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547839 Supplier Invitrogen™ Supplier No. PA547839
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

GDF-15 belongs to the TGF-beta cytokine family, whose members play an important role during prenatal development and postnatal growth, and the remodeling and maintenance of a variety of tissues and organs. GDF-15 is expressed predominantly in the placenta and, to a much lesser extent, in various other tissues. The presence of GDF-15 in amniotic fluid and its elevated levels in the sera of pregnant women suggest GDF-15's involvement in gestation and embryonic development. GDF-15 generally exerts tumor suppressive activities and is one of the predominant factors produced and secreted in response to activation of the p53 pathway. Interestingly, the serum level of GDF-15 is positively correlated with neoplastic progression of several tumor types, including certain colorectal, pancreatic, and prostate cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF15
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Gdf15
Gene Accession No. Q9Z0J7
Gene Alias Gdf15; GDF-15; growth differentiation factor 15; growth/differentiation factor 15; macrophage inhibiting cytokine-1; macrophage inhibitory cytokine 1; MIC1; MIC-1; NAG1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; PDF; PLAB; Placental bone morphogenetic protein; placental TGF-beta; Prostate differentiation factor; PTGFB; PTGF-beta; SBF
Gene Symbols Gdf15
Host Species Sheep
Immunogen E. coli-derived recombinant mouse GDF-15 Ser189-Ala303.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23886
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.