Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735672 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5672 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5672 Supplier Proteintech Group Inc Supplier No. 274551AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bone morphogenetic proteins (e. g., BMP5, MIM 112265) are members of the transforming growth factor-beta (see TGFB1, MIM 190180) superfamily and regulate tissue differentiation and maintece. They are synthesized as precursor molecules that are processed at a dibasic cleavage site to release C-terminal domains containing a characteristic motif of 7 conserved cysteines in the mature protein.

Specifications

Antigen GDF15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GDF15
Gene Accession No. Q99988, Q9Z0J7
Gene Alias GDF 15, GDF15, MIC 1, MIC1, NAG 1, NRG 1, NSAID activated gene 1 protein, NSAID regulated gene 1 protein, PDF, PLAB, Placental TGF beta, PTGFB
Gene Symbols GDF15
Host Species Rabbit
Immunogen GDF15 Fusion Protein Ag26760
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23886, 9518
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.