Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDF15 Recombinant Rabbit Monoclonal Antibody (10K4Y8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551358
Description
Immunogen sequence: HLHLRISRAA LPEGLPEASR LHRALFRLSP TASRSWDVTR PLRRQLSLAR PQAPALHLRL SPPPSQSDQL LAESSSARPQ LELHLRPQAA RGRRRARARN G.
GDF-15 belongs to the TGF-beta cytokine family, whose members play an important role during prenatal development and postnatal growth, and the remodeling and maintenance of a variety of tissues and organs. GDF-15 is expressed predominantly in the placenta and, to a much lesser extent, in various other tissues. The presence of GDF-15 in amniotic fluid and its elevated levels in the sera of pregnant women suggest GDF-15's involvement in gestation and embryonic development. GDF-15 generally exerts tumor suppressive activities and is one of the predominant factors produced and secreted in response to activation of the p53 pathway. Interestingly, the serum level of GDF-15 is positively correlated with neoplastic progression of several tumor types, including certain colorectal, pancreatic, and prostate cancers.
Specifications
GDF15 | |
Recombinant Monoclonal | |
0.2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q99988 | |
Gdf15 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human GDF15 (NP_004855.2). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
ELISA, Western Blot | |
10K4Y8 | |
Unconjugated | |
Gdf15 | |
Gdf15; GDF-15; growth differentiation factor 15; growth/differentiation factor 15; macrophage inhibiting cytokine-1; macrophage inhibitory cytokine 1; MIC1; MIC-1; NAG1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; PDF; PLAB; Placental bone morphogenetic protein; placental TGF-beta; Prostate differentiation factor; PTGFB; PTGF-beta; SBF | |
Rabbit | |
Affinity chromatography | |
RUO | |
9518 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction