Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDF5OS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584562 Supplier Invitrogen™ Supplier No. PA584562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPSFSRALM SNTYLCFLTT GPRSSRENTQ KSFPATSPRE Highest antigen sequence indentity to the following orthologs: Rat - 35%, Mouse - 85%.

GDF5OS is a protein coding gene. Gene ontology (GO) annotation include mitochondrion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF5OS
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GDF5OS
Gene Accession No. Q5U4N7
Gene Alias GDF5OS; growth differentiation factor 5 opposite strand; Protein GDF5OS
Gene Symbols GDF5OS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GDF5OS. Recombinant protein control fragment (Product #RP-101610).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 554250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.