Learn More
Invitrogen™ GDF7 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547941
Description
Approximately 10% cross-reactivity with recombinant mouse (rm) GDF-5 and rmGDF-6 is observed and less than 1% cross-reactivity with rmGDF-1, rmGDF-3, rmGDF-8, rmGDF-9, and recombinant human GDF-11 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord.
Specifications
GDF7 | |
Polyclonal | |
Unconjugated | |
GDF7 | |
BMP12; Gdf7; Gdf-7; growth differentiation factor 7; growth/differentiation factor 7 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
238057 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P43029 | |
GDF7 | |
E. coli-derived recombinant mouse GDF-7 Thr316-Arg461. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.