Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564403 Supplier Invitrogen™ Supplier No. PA564403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YNINNSVSFS SAVKCVCNLM IKEPKSSSRT LGRAPYSFTF NSQFEFGKKH KWIQIDVTSL LQPLVASNKR SIHMSINFTC MKDQLEHP Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 58%.

GDF9 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues.Growth factors synthesized by ovarian somatic cells directly affect oocyte growth and function. GDF9 is expressed in oocytes and is thought to be required for ovarian folliculogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GDF9
Gene Accession No. O60383
Gene Alias Gdf9; Gdf-9; growth differentiation factor 9; growth/differentiation factor 9
Gene Symbols GDF9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GDF9. Recombinant protein control fragment (Product #RP-104085).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.