Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF9 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA547924
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547924 Supplier Thermo Scientific Supplier No. PA547924

Goat Polyclonal Antibody

This antibody detects mouse GDF-9 in direct ELISAs and both mouse and hamster GDF-9 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) GDF-1, rmGDF-5, rmGDF-6, rmGDF-8, and rmGDF-15 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

GDF9 Polyclonal antibody specifically detects GDF9 in Hamster, Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF9
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene GDF9
Gene Accession No. Q07105
Gene Alias Gdf9; Gdf-9; growth differentiation factor 9; growth/differentiation factor 9
Gene Symbols GDF9
Host Species Goat
Immunogen E. coli-derived recombinant mouse GDF-9
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100756618, 14566
Target Species Hamster, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.