Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDI1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299205AF488
Description
GDI1 Polyclonal specifically detects GDI1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GDI1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
FLJ41411, GDI-1, GDIL, GDP dissociation inhibitor 1, Guanosine diphosphate dissociation inhibitor 1, mental retardation, X-linked 41, mental retardation, X-linked 48, MRX41, oligophrenin-2, OPHN2MRX48, Protein XAP-4, Rab GDI alpha, rab GDP dissociation inhibitor alpha, rab GDP-dissociation inhibitor, alpha, RABGDIARABGD1A, XAP4, XAP-4 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human GDI1. | |
0.1 mL | |
Signal Transduction | |
2664 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction