Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581504 Supplier Invitrogen™ Supplier No. PA581504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human GDI1.

GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI1 is expressed primarily in neural and sensory tissues. Mutations in GDI1 have been linked to X-linked nonspecific mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDI1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GDI1
Gene Accession No. P31150
Gene Alias 1A; GDI1; GDI-1; GDIA; GDIalpha; GDIL; GDP dissociation inhibitor 1; GDP-dissociation inhibitor 1; GTP-binding protein; guanosine diphosphate (GDP) dissociation inhibitor 1; guanosine diphosphate dissociation inhibitor 1; mental retardation, X-linked 48; MRX41; MRX48; oligophrenin-2; OPHN2; Protein XAP-4; rab GDI alpha; Rab GDIalpha; rab GDP dissociation inhibitor alpha; rab GDP-dissociation inhibitor, alpha; RABGD RABGDIA; RABGD1A; RABGDIA; SMG p25A GDI; testis secretory sperm-binding protein Li 208a; XAP4; XAP-4
Gene Symbols GDI1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Human GDI1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.