Learn More
GDI2 Mouse anti-Human, Mouse, Clone: 4B8F3, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 600781IG150UL

Description
This antibody can bind both GDIs for the close sequences.
Regulates the GDP/GTP exchange reaction of most Rab proteins by inhibiting the dissociation of GDP from them, and the subsequent binding of GTP to them.Specifications
GDI2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50395, Q61598 | |
Gdi2 | |
GDI2 Fusion Protein Ag4613 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
4B8F3 | |
Unconjugated | |
GDI2 | |
GDI 2, GDI2, GDP dissociation inhibitor 2, Rab GDI beta, RABGDIB | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
14569, 2665 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.