Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599998 Supplier Invitrogen™ Supplier No. PA599998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GDI2.

GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI2 is ubiquitously expressed. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDI2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GDI2
Gene Accession No. P50395, P50399, Q61598
Gene Alias epididymis secretory sperm binding protein Li 46e; GDI beta; Gdi2; GDI-2; Gdi3; GDI-3; GDIB; GDI-B; GDP dissociation inhibitor 2; guanosine diphosphate (GDP) dissociation inhibitor 2; guanosine diphosphate (GDP) dissociation inhibitor 3; guanosine diphosphate dissociation inhibitor 2; guanosine diphosphate dissociation inhibitor 3; HEL-S-46e; LOW QUALITY PROTEIN: rab GDP dissociation inhibitor beta; rab GDI beta; Rab GDP dissociation inhibitor beta; RAB GDP dissociation inhibitor beta-2; rab GDP dissociation inhibitor beta-like protein; rab GDP-dissociation inhibitor, beta; RABGDIB
Gene Symbols GDI2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GDI2(Accession P50395), corresponding to amino acid residues E292-N342.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14569, 2665, 29662
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.