Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548631
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548631 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548631 Supplier Invitrogen™ Supplier No. PA548631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GDI2 Polyclonal Antibody for Western Blot, IHC (P)

GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI2 is ubiquitously expressed. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDI2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GDI2
Gene Accession No. P50395
Gene Alias epididymis secretory sperm binding protein Li 46e; GDI beta; Gdi2; GDI-2; Gdi3; GDI-3; GDIB; GDI-B; GDP dissociation inhibitor 2; guanosine diphosphate (GDP) dissociation inhibitor 2; guanosine diphosphate (GDP) dissociation inhibitor 3; guanosine diphosphate dissociation inhibitor 2; guanosine diphosphate dissociation inhibitor 3; HEL-S-46e; LOW QUALITY PROTEIN: rab GDP dissociation inhibitor beta; rab GDI beta; Rab GDP dissociation inhibitor beta; RAB GDP dissociation inhibitor beta-2; rab GDP dissociation inhibitor beta-like protein; rab GDP-dissociation inhibitor, beta; RABGDIB
Gene Symbols GDI2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 135-164 amino acids from the Central region of human GDI2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2665
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.