Learn More
Invitrogen™ GDI2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597133
Description
Immunogen sequence: QVIRVICILS HPIKNTNDAN SCQIIIPQNQ VNRKSDIYVC MISFAHNVAA QGKYIAIVST TVETKEPEKE IRPALELLEP IEQKFVSISD LLVPKDLGTE SQIFISRTYD ATTHFETTCD DIKNIYKRMT GSEFDFEEMK RKKNDIYGED; Positive Samples: HT-29, A-549, HepG2, U-87MG, Mouse spleen, Mouse kidney, Mouse lung, Rat liver, Rat spinal cord; Cellular Location: Cytoplasm, Membrane, Peripheral membrane protein.
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI2 is ubiquitously expressed. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements.
Specifications
GDI2 | |
Polyclonal | |
Unconjugated | |
GDI2 | |
epididymis secretory sperm binding protein Li 46e; GDI beta; Gdi2; GDI-2; Gdi3; GDI-3; GDIB; GDI-B; GDP dissociation inhibitor 2; guanosine diphosphate (GDP) dissociation inhibitor 2; guanosine diphosphate (GDP) dissociation inhibitor 3; guanosine diphosphate dissociation inhibitor 2; guanosine diphosphate dissociation inhibitor 3; HEL-S-46e; LOW QUALITY PROTEIN: rab GDP dissociation inhibitor beta; rab GDI beta; Rab GDP dissociation inhibitor beta; RAB GDP dissociation inhibitor beta-2; rab GDP dissociation inhibitor beta-like protein; rab GDP-dissociation inhibitor, beta; RABGDIB | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14569, 2665, 29662 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P50395, P50399, Q61598 | |
GDI2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 296-445 of human GDI2 (NP_0014852). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.