Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDN Polyclonal Antibody, Cyanine5, Bioss
SDP

Catalog No. 502281992
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1992 Supplier Bioss Supplier No. BS9845RCY5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GDN Polyclonal antibody specifically detects GDN in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen GDN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine5
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene SERPINE2
Gene Accession No. P07092, P07093, Q07235
Gene Alias B230326M24Rik; EGK_04851; GDN; GDNPF; Gdnpn1; Glia derived nexin; glia-derived nexin; Glia-derived nexin/protease nexin I; Glia-derived nexin-like protein; glial-derived neurite promoting factor; P17; PAI-1; Peptidase inhibitor 7; Pi7; PI-7; plasminogen activator inhibitor type 1; PN1; Pn-1; PNI; Protease nexin 1; protease nexin I; protease nexin PN-1; protease nexin-1; serine (or cysteine) peptidase inhibitor, clade E, member 2; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serine (or cysteine) proteinase inhibitor, clade E, member 2; serine protease inhibitor 4; serine protease-inhibitor 4; Serpin E2; serpin family E member 2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serpin peptidase inhibitor, clade E, member 2; SERPINE 2; SERPINE1; Serpine2; Spi4; Spin4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GDN/SERPINE2
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20720, 29366, 5270
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.