Learn More
Invitrogen™ GDNF Recombinant Rabbit Monoclonal Antibody (11D2)

Description
GDNF Recombinant Monoclonal Antibody for ELISA, Flow, IHC (P), IP, Western Blot
Specifications
Specifications
| Antigen | GDNF |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 11D2 |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | GDNF |
| Gene Accession No. | P39905 |
| Gene Alias | AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.